Assay Details
| Gene Symbol: Mxra7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026280.3 | NP_080556.1 | 2 - 3 | 372 | 64 | |
| GenBank mRNA | AK012601.1 | - | 2 - 3 | 344 | 64 | |
| AK075801.1 | - | 2 - 3 | 373 | 64 | ||
| BC075618.1 | - | 2 - 3 | 339 | 64 | ||
| BC147320.1 | - | 2 - 3 | 453 | 9056373 | 64 | |
| BC147321.1 | - | 2 - 3 | 453 | 9056376 | 64 | |
More Information
Set Membership: |
|

Mxra7
Genomic Map