Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200044876.mm.1 |
NM_001164369.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044876.mm.1 |
NM_001164369.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044876.mm.1 |
NM_029815.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044876.mm.1 |
NM_029815.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044876.mm.1 |
TC0200005249.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200044877.mm.1 |
NM_001164369.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044877.mm.1 |
NM_001164369.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044877.mm.1 |
NM_029815.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044877.mm.1 |
NM_029815.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044877.mm.1 |
TC0200005249.mm.1 |
MTA TC |