Assay Details
| Gene Symbol: Gfap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010277.3 | NP_034407.2 | 8 - 9 | 1327 | 68 | |
| GenBank mRNA | AF332061.1 | - | 8 - 9 | 1154 | 68 | |
| AF332062.1 | - | 8 - 9 | 1154 | 68 | ||
| AK079577.1 | - | 8 - 9 | 1262 | 68 | ||
| BC100728.1 | - | 8 - 9 | 1206 | 68 | ||
| BC100737.1 | - | 8 - 9 | 1199 | 68 | ||
| BC101968.1 | - | 8 - 9 | 1206 | 68 | ||
| BC103571.1 | - | 8 - 9 | 1200 | 68 | ||
| BC139357.1 | - | 8 - 9 | 1327 | 8862379 | 68 | |
| BC139358.1 | - | 8 - 9 | 1327 | 8862380 | 68 | |
| K01347.1 | - | 8 - 9 | 1163 | 68 | ||
| M25937.1 | - | 8 - 9 | 1163 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gfap
Genomic Map