Assay Details
| Gene Symbol: Smad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008539.3 | NP_032565.2 | 2 - 3 | 801 | 71 | |
| GenBank mRNA | AK017583.1 | - | 2 - 3 | 762 | 71 | |
| AK034045.1 | - | 2 - 3 | 762 | 71 | ||
| AK054104.1 | - | 2 - 3 | 762 | 71 | ||
| AK084505.1 | - | 2 - 3 | 869 | 71 | ||
| BC058693.1 | - | 2 - 3 | 801 | 6811514 | 71 | |
| U74359.1 | - | 2 - 3 | 667 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smad1
Genomic Map