Assay Details
| Gene Symbol: Slc10a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009981.2 | NP_001009981.1 | 4 - 5 | 642 | 91 | |
| NM_001282108.1 | NP_001269037.1 | 4 - 5 | 642 | 91 | ||
| NM_029736.2 | NP_084012.1 | 4 - 5 | 642 | 91 | ||
| NR_104094.1 | 3 - 4 | 559 | 91 | |||
| GenBank mRNA | AK010834.1 | - | 4 - 5 | 592 | 91 | |
| AK040517.1 | - | 4 - 5 | 521 | 91 | ||
| AK079901.1 | - | 4 - 5 | 619 | 91 | ||
| AK152752.1 | - | 4 - 5 | 603 | 91 | ||
| AK152849.1 | - | 4 - 5 | 603 | 91 | ||
| AK160402.1 | - | 4 - 5 | 626 | 91 | ||
| AY825926.1 | - | 4 - 5 | 395 | 91 | ||
| AY825927.1 | - | 3 - 4 | 312 | 91 | ||
| AY825928.1 | - | 4 - 5 | 395 | 91 | ||
| BC116666.1 | - | 4 - 5 | 508 | 40107077 | 91 | |
| BC116717.1 | - | 4 - 5 | 508 | 40107078 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc10a7
Genomic Map