Assay Details
| Gene Symbol: Slc10a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282108.1 | NP_001269037.1 | 7 - 8 | 804 | 63 | |
| NM_001282109.1 | NP_001269038.1 | 6 - 7 | 728 | 63 | ||
| NM_029736.2 | NP_084012.1 | 7 - 8 | 804 | 63 | ||
| NR_104094.1 | 6 - 7 | 721 | 63 | |||
| GenBank mRNA | AK010834.1 | - | 7 - 8 | 754 | 63 | |
| AK040517.1 | - | 7 - 8 | 683 | 63 | ||
| AK051650.1 | - | 4 - 5 | 566 | 63 | ||
| AK079901.1 | - | 7 - 8 | 781 | 63 | ||
| AK152752.1 | - | 7 - 8 | 765 | 63 | ||
| AK152849.1 | - | 7 - 8 | 765 | 63 | ||
| AK160402.1 | - | 7 - 8 | 788 | 63 | ||
| AK165983.1 | - | 6 - 7 | 665 | 63 | ||
| AY825927.1 | - | 6 - 7 | 474 | 63 | ||
| BC116666.1 | - | 7 - 8 | 670 | 40107077 | 63 | |
| BC116717.1 | - | 7 - 8 | 670 | 40107078 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc10a7
Genomic Map