Assay Details
Gene Symbol: Hsp90b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011631.1 | NP_035761.1 | 12 - 13 | 1733 | 135 | |
GenBank mRNA | AK031918.1 | - | 8 - 9 | 1085 | 135 | |
AK146885.1 | - | 12 - 13 | 1799 | 135 | ||
AK150811.1 | - | 12 - 13 | 1753 | 135 | ||
AK151413.1 | - | 12 - 13 | 1750 | 135 | ||
AK153550.1 | - | 12 - 13 | 1753 | 135 | ||
AK159974.1 | - | 12 - 13 | 1758 | 135 | ||
AK160093.1 | - | 12 - 13 | 1752 | 135 | ||
AK160827.1 | - | 1173 | 135 | |||
AK163243.1 | - | 2 - 3 | 311 | 135 | ||
AK165975.1 | - | 12 - 13 | 1753 | 135 | ||
AK168952.1 | - | 12 - 13 | 1753 | 135 | ||
AK169090.1 | - | 12 - 13 | 1753 | 135 | ||
BC010445.1 | - | 12 - 13 | 1665 | 3587178 | 135 | |
BC011439.1 | - | 12 - 13 | 1739 | 3707817 | 135 | |
J03297.1 | - | 12 - 13 | 1733 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
