Assay Details
| Gene Symbol: Hsp90b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011631.1 | NP_035761.1 | 6 - 7 | 940 | 188 | |
| GenBank mRNA | AK031918.1 | - | 2 - 3 | 292 | 188 | |
| AK088148.1 | - | 4 - 5 | 643 | 188 | ||
| AK146885.1 | - | 6 - 7 | 1006 | 188 | ||
| AK150811.1 | - | 6 - 7 | 960 | 188 | ||
| AK151413.1 | - | 6 - 7 | 957 | 188 | ||
| AK153550.1 | - | 6 - 7 | 960 | 188 | ||
| AK159974.1 | - | 6 - 7 | 965 | 188 | ||
| AK160093.1 | - | 6 - 7 | 959 | 188 | ||
| AK160827.1 | - | 380 | 188 | |||
| AK165975.1 | - | 6 - 7 | 960 | 188 | ||
| AK168952.1 | - | 6 - 7 | 960 | 188 | ||
| AK169090.1 | - | 6 - 7 | 960 | 188 | ||
| BC010445.1 | - | 6 - 7 | 872 | 3587178 | 188 | |
| BC011439.1 | - | 6 - 7 | 946 | 3707817 | 188 | |
| J03297.1 | - | 6 - 7 | 940 | 188 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsp90b1
Genomic Map