Assay Details
| Gene Symbol: Hsp90b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011631.1 | NP_035761.1 | 7 - 8 | 1065 | 198 | |
| GenBank mRNA | AK031918.1 | - | 3 - 4 | 417 | 198 | |
| AK088148.1 | - | 5 - 6 | 768 | 198 | ||
| AK146885.1 | - | 7 - 8 | 1131 | 198 | ||
| AK150811.1 | - | 7 - 8 | 1085 | 198 | ||
| AK151413.1 | - | 7 - 8 | 1082 | 198 | ||
| AK153550.1 | - | 7 - 8 | 1085 | 198 | ||
| AK159974.1 | - | 7 - 8 | 1090 | 198 | ||
| AK160093.1 | - | 7 - 8 | 1084 | 198 | ||
| AK160827.1 | - | 505 | 198 | |||
| AK165975.1 | - | 7 - 8 | 1085 | 198 | ||
| AK168952.1 | - | 7 - 8 | 1085 | 198 | ||
| AK169090.1 | - | 7 - 8 | 1085 | 198 | ||
| BC010445.1 | - | 7 - 8 | 997 | 3587178 | 198 | |
| BC011439.1 | - | 7 - 8 | 1071 | 3707817 | 198 | |
| J03297.1 | - | 7 - 8 | 1065 | 198 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsp90b1
Genomic Map