Assay Details
| Gene Symbol: Hsp90b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011631.1 | NP_035761.1 | 9 - 10 | 1323 | 104 | |
| GenBank mRNA | AK031918.1 | - | 5 - 6 | 675 | 104 | |
| AK146885.1 | - | 9 - 10 | 1389 | 104 | ||
| AK150811.1 | - | 9 - 10 | 1343 | 104 | ||
| AK151413.1 | - | 9 - 10 | 1340 | 104 | ||
| AK153550.1 | - | 9 - 10 | 1343 | 104 | ||
| AK159974.1 | - | 9 - 10 | 1348 | 104 | ||
| AK160093.1 | - | 9 - 10 | 1342 | 104 | ||
| AK160827.1 | - | 763 | 104 | |||
| AK165975.1 | - | 9 - 10 | 1343 | 104 | ||
| AK168952.1 | - | 9 - 10 | 1343 | 104 | ||
| AK169090.1 | - | 9 - 10 | 1343 | 104 | ||
| BC010445.1 | - | 9 - 10 | 1255 | 3587178 | 104 | |
| BC011439.1 | - | 9 - 10 | 1329 | 3707817 | 104 | |
| J03297.1 | - | 9 - 10 | 1323 | 104 | ||
| M29652.1 | - | 1 - 2 | 103 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsp90b1
Genomic Map