Assay Details
| Gene Symbol: Rab4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009003.3 | NP_033029.2 | 4 - 5 | 433 | 114 | |
| GenBank mRNA | AB232591.1 | - | 4 - 5 | 293 | 114 | |
| AK136566.1 | - | 4 - 5 | 422 | 114 | ||
| AK145452.1 | - | 4 - 5 | 416 | 114 | ||
| AK161877.1 | - | 4 - 5 | 401 | 114 | ||
| BC138508.1 | - | 4 - 5 | 346 | 8861528 | 114 | |
| CO806247.1 | - | 449 | 114 | |||
| D86563.1 | - | 4 - 5 | 407 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab4a
Genomic Map