Assay Details
| Gene Symbol: Pde6g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF190928.1 | - | 2 - 3 | 185 | 103 | |
| AK080735.1 | - | 3 - 4 | 331 | 103 | ||
| BC082288.1 | - | 3 - 4 | 288 | 6490841 | 103 | |
| Y00746.1 | - | 3 - 4 | 306 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2177864 | AK080735.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2177864 | NM_012065.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137842 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 278914 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 374485 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 88912 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 61781 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115257 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2177864 | AK080735.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2177864 | NM_012065.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P398974 | BC014723.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P398974 | BC082288.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P398974 | AK080735.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P398974 | NM_012065.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184641 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184640 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115257 | - | - |
| Whole Mouse Genome G4122A | A_52_P398974 | BC014723.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P398974 | BC082288.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P398974 | AK080735.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P398974 | NM_012065.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139752 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115257 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09373289 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09373296 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09373290 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09373297 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09373301 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09373299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184653 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09373290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09147567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09373286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09147570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09147579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09147571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09447283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09147572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09373301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09447281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09373294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184646 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10393799 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393796 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393795 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393800 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393794 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425100_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450453_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10393793 | - | - |
| Mouse Genome 430 2.0 Array | 1425100_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1425100_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1450453_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1450453_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18588_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18588_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100040102.mm.1 | NM_012065.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040102.mm.1 | TC1100004206.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425100_a_at | - | - |
| Mouse Expression Set 430 | 1450453_a_at | - | - |
| Mouse Expression Set 430 | 1425100_PM_a_at | - | - |
| Mouse Expression Set 430 | 1450453_PM_a_at | - | - |
| Murine Genome U74v2 Set | 97732_at | - | - |
| Murine Genome U74v2 Set | 97732_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450453_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450453_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425100_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425100_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450453_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450453_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425100_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425100_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 60102 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 60102 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000060102 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2590508 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2590508 | - | - |
| mouse-6 v1.1 (Illumina) | scl39239.4.1_19-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 60102 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39239.4.1_19-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 60102 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2590508 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2590508 | - | - |

Pde6g
Genomic Map