Assay Details
Gene Symbol: Notum | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175263.4 | NP_780472.3 | 3 - 4 | 473 | 58 | |
GenBank mRNA | AK019987.1 | - | 3 - 4 | 473 | 58 | |
DN174397.1 | - | 3 - 4 | 479 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
