Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100040089.mm.1 |
NM_001195023.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040089.mm.1 |
NM_001195023.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040089.mm.1 |
NM_199469.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040089.mm.1 |
NM_199469.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040089.mm.1 |
TC1100004204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040090.mm.1 |
NM_001195023.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040090.mm.1 |
NM_001195023.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040090.mm.1 |
NM_199469.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040090.mm.1 |
NM_199469.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040090.mm.1 |
TC1100004204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040086.mm.1 |
NM_001195023.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040086.mm.1 |
NM_001195023.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040086.mm.1 |
NM_199469.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040086.mm.1 |
NM_199469.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040086.mm.1 |
TC1100004204.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100040087.mm.1 |
NM_001195023.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040087.mm.1 |
NM_001195023.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040087.mm.1 |
NM_199469.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040087.mm.1 |
NM_199469.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100040087.mm.1 |
TC1100004204.mm.1 |
MTA TC |