Assay Details
| Gene Symbol: Arhgdia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK077343.1 | - | 1 - 2 | 257 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK077343.1 transcripts.
Set Membership: |
|

Arhgdia
Genomic Map