Assay Details
| Gene Symbol: Mettl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026197.3 | NP_080473.1 | 5 - 6 | 712 | 83 | |
| GenBank mRNA | AK011780.1 | - | 5 - 6 | 713 | 83 | |
| AK012739.1 | - | 5 - 6 | 713 | 83 | ||
| AK013306.1 | - | 5 - 6 | 724 | 83 | ||
| AK029045.1 | - | 5 - 6 | 696 | 83 | ||
| AK145022.1 | - | 5 - 6 | 720 | 83 | ||
| AK145662.1 | - | 5 - 6 | 712 | 83 | ||
| AK168798.1 | - | 5 - 6 | 662 | 83 | ||
| BC003467.1 | - | 5 - 6 | 683 | 83 | ||
| BC022902.1 | - | 5 - 6 | 724 | 83 | ||
| BC049897.1 | - | 5 - 6 | 641 | 3979938 | 83 | |
| BC054724.1 | - | 5 - 6 | 648 | 5695738 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl16
Genomic Map