Assay Details
| Gene Symbol: Dph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144491.2 | NP_652762.2 | 5 - 6 | 582 | 82 | |
| GenBank mRNA | AK082812.1 | - | 5 - 6 | 587 | 82 | |
| AK128996.1 | - | 573 | 82 | |||
| AY078170.1 | - | 5 - 6 | 573 | 82 | ||
| BC044663.1 | - | 5 - 6 | 553 | 82 | ||
| BC080761.1 | - | 5 - 6 | 558 | 82 | ||
More Information
Set Membership: |
|

Dph1
Genomic Map