Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900000165.mm.1 |
NM_001033322.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000165.mm.1 |
NM_001033322.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000165.mm.1 |
TC0900000019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000167.mm.1 |
NM_001033322.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000167.mm.1 |
NM_001033322.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000167.mm.1 |
TC0900000019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000172.mm.1 |
NM_001033322.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000172.mm.1 |
NM_001033322.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000172.mm.1 |
TC0900000019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000174.mm.1 |
NM_001033322.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000174.mm.1 |
NM_001033322.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000174.mm.1 |
TC0900000019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000169.mm.1 |
NM_001033322.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000169.mm.1 |
NM_001033322.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000169.mm.1 |
TC0900000019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000171.mm.1 |
NM_001033322.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000171.mm.1 |
NM_001033322.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000171.mm.1 |
TC0900000019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900000166.mm.1 |
NM_001033322.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000166.mm.1 |
NM_001033322.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900000166.mm.1 |
TC0900000019.mm.1 |
MTA TC |