Assay Details
| Gene Symbol: Vps4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126165.1 | NP_569053.1 | 7 - 8 | 915 | 84 | |
| GenBank mRNA | AK047821.1 | - | 7 - 8 | 927 | 84 | |
| AK159117.1 | - | 7 - 8 | 941 | 84 | ||
| AK169949.1 | - | 7 - 8 | 903 | 84 | ||
| BC018368.1 | - | 7 - 8 | 915 | 3963907 | 84 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Vps4a
Genomic Map