Assay Details
Gene Symbol: Impa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053261.2 | NP_444491.1 | 5 - 6 | 674 | 57 | |
GenBank mRNA | AF353730.1 | - | 5 - 6 | 673 | 57 | |
AK154489.1 | - | 5 - 6 | 712 | 57 | ||
AK154845.1 | - | 5 - 6 | 645 | 57 | ||
AK170962.1 | - | 5 - 6 | 712 | 57 | ||
AK171626.1 | - | 5 - 6 | 647 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
