Assay Details
| Gene Symbol: Gfra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010279.3 | NP_034409.1 | 5 - 6 | 878 | 66 | |
| NR_104342.1 | 5 - 6 | 878 | 66 | |||
| GenBank mRNA | AB000800.1 | - | 4 - 5 | 432 | 66 | |
| AK076171.1 | - | 5 - 6 | 798 | 66 | ||
| BC044783.2 | - | 5 - 6 | 757 | 66 | ||
| BC054378.1 | - | 4 - 5 | 549 | 6390018 | 66 | |
| KM079499.1 | - | 4 - 5 | 537 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gfra1
Genomic Map