Assay Details
Gene Symbol: C8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290645.1 | NP_001277574.1 | 4 - 5 | 605 | 128 | |
NM_146148.2 | NP_666260.1 | 5 - 6 | 702 | 128 | ||
GenBank mRNA | AB077295.1 | - | 5 - 6 | 716 | 128 | |
AB077296.1 | - | 5 - 6 | 665 | 128 | ||
AK090245.1 | - | 5 - 6 | 700 | 129 | ||
BC027748.1 | - | 5 - 6 | 698 | 5052412 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
