Assay Details
Gene Symbol: 1700024P16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162980.1 | NP_001156452.1 | 16 - 17 | 2116 | 154 | |
NM_001285952.1 | NP_001272881.1 | 17 - 18 | 2194 | 154 | ||
NM_001285953.1 | NP_001272882.1 | 16 - 17 | 2049 | 154 | ||
NR_104376.1 | 17 - 18 | 2155 | 198 | |||
GenBank mRNA | AK028077.1 | - | 16 - 17 | 2056 | 154 | |
AK133762.1 | - | 16 - 17 | 1987 | 154 | ||
BC137723.1 | - | 16 - 17 | 2116 | 8860742 | 154 | |
BC144857.1 | - | 17 - 18 | 2194 | 9053405 | 154 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2116689 | AK133762.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2116689 | AV174094.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2116689 | NM_001162980.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984881 | NM_001162980.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641761 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 354402 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425269 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371570 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 186286 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 965808 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96401 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 28480 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 289893 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 71915 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 850618 | AK038305.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 228075 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 285049 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477118 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 262471 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544784 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 912123 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 926359 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 93020 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 744359 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 566803 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 68317 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 439514 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 321769 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 869079 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 165857 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932904 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 721863 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 455193 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 104677 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 64120 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 272012 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 513547 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527149 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 349021 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655082 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 166261 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 256203 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 26030 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 657063 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 589688 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 619436 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 349128 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 847149 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593657 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 954140 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779290 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 383267 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 681585 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 809939 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 695141 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740775 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 154359 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 362299 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 605966 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392336 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 160621 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939591 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704998 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 791557 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 754169 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 89520 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 34834 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52893 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 66351 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 93252 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 53410 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 130008 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 80096 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 35057 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110410 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165529 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2116689 | AK133762.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2116689 | AV174094.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2116689 | NM_001162980.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984881 | NM_001162980.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05345071 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05345230 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05345150 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05345134 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110410 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05345036 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05345012 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00950119 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165529 | - | - |
Whole Mouse Genome G4122A | A_52_P319552 | BC144858.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P319552 | BC144857.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P319552 | AV174094.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P319552 | BC137723.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P319552 | NM_001162980.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05345036 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00950119 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110410 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165529 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175792 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175927 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03971434 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03971441 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03477415 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175933 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03588909 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03638640 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03324872 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03638648 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03638646 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175769 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03211935 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175680 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175699 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03211928 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175839 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175684 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175709 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175708 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175783 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175626 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175799 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03588902 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175763 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03469022 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03839262 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175676 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03587431 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03393342 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175647 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175692 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03965035 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175836 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175849 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03638641 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175621 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03643067 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03653695 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03211947 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175642 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175782 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03588914 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175681 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175786 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03858431 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03377543 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175694 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175887 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175926 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03638633 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04058737 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03685669 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175635 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175777 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175833 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04077863 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175657 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175776 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175779 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175678 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03258966 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175766 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175618 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03388935 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175689 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175874 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175850 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175879 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03965034 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175625 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175641 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175925 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03638642 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175634 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03738158 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175658 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175637 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175846 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03965041 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175630 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175696 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03500756 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175830 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03638647 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175774 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175931 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175800 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03505183 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03762951 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175639 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03477421 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03653693 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175787 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175825 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03588904 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03588918 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03850532 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03245603 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03651756 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175806 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175764 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175794 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175687 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175841 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175653 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175655 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175646 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175873 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175648 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175785 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175826 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03388933 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175847 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175620 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03971428 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03706441 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175644 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03391306 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175631 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175682 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175918 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03377542 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175818 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175827 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03211938 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03638631 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175924 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175697 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03505191 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03211943 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03211926 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175762 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23175670 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10506484 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506474 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506478 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506479 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506481 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506485 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506476 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506477 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506472 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506473 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506482 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506487 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506471 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506475 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506483 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506480 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10506486 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10506470 | - | - |
Affymetrix Mouse Clariom S Array | PSR0400007460.mm.1 | NM_001162980.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007460.mm.1 | NM_001162980.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007460.mm.1 | NM_001285952.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007460.mm.1 | NM_001285953.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007460.mm.1 | NR_104376.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007460.mm.1 | TC0400001090.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400007461.mm.1 | NM_001162980.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007461.mm.1 | NM_001162980.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007461.mm.1 | NM_001285952.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007461.mm.1 | NM_001285953.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007461.mm.1 | NR_104376.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007461.mm.1 | TC0400001090.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400007463.mm.1 | NM_001162980.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007463.mm.1 | NM_001162980.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007463.mm.1 | NM_001285952.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007463.mm.1 | NM_001285953.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007463.mm.1 | NR_104376.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007463.mm.1 | TC0400001090.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400007459.mm.1 | NM_001162980.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007459.mm.1 | NM_001162980.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007459.mm.1 | NM_001285952.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007459.mm.1 | NM_001285953.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007459.mm.1 | NR_104376.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007459.mm.1 | TC0400001090.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400007462.mm.1 | NM_001162980.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007462.mm.1 | NM_001162980.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007462.mm.1 | NM_001285952.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007462.mm.1 | NM_001285953.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007462.mm.1 | NR_104376.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400007462.mm.1 | TC0400001090.mm.1 | MTA TC |
Murine Genome U74v2 Set | 166704_PM_i_at | - | - |
Murine Genome U74v2 Set | 166704_i_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 501485 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 870706 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2739065 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2739065 | - | - |
