Assay Details
| Gene Symbol: 1700024P16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162980.1 | NP_001156452.1 | 10 - 11 | 1722 | 92 | |
| NM_001285952.1 | NP_001272881.1 | 11 - 12 | 1800 | 92 | ||
| NM_001285953.1 | NP_001272882.1 | 10 - 11 | 1655 | 92 | ||
| NR_104376.1 | 10 - 11 | 1722 | 92 | |||
| GenBank mRNA | AK028077.1 | - | 10 - 11 | 1662 | 92 | |
| AK133762.1 | - | 10 - 11 | 1593 | 92 | ||
| BC137723.1 | - | 10 - 11 | 1722 | 8860742 | 92 | |
| BC144857.1 | - | 11 - 12 | 1800 | 9053405 | 92 | |
| BC144858.1 | - | 10 - 11 | 1723 | 9053406 | 92 | |
More Information
Set Membership: |
|

1700024P16Rik
Genomic Map