Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100003158.mm.1 |
NM_001114663.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003158.mm.1 |
NM_001114663.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003158.mm.1 |
TC0100000404.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100003160.mm.1 |
NM_001114663.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003160.mm.1 |
NM_001114663.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003160.mm.1 |
TC0100000404.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100003159.mm.1 |
NM_001114663.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003159.mm.1 |
NM_001114663.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003159.mm.1 |
TC0100000404.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100003157.mm.1 |
NM_001114663.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003157.mm.1 |
NM_001114663.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003157.mm.1 |
TC0100000404.mm.1 |
MTA TC |