Assay Details
| Gene Symbol: Plcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114663.1 | NP_001108135.1 | 3 - 4 | 3362 | 63 | |
| GenBank mRNA | AI644888.1 | - | 1 - 2 | 171 | 63 | |
| AK048715.1 | - | 1062 | 63 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Plcl1
Genomic Map