Assay Details
Gene Symbol: Mrps9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023514.4 | NP_076003.3 | 2 - 3 | 372 | 74 | |
GenBank mRNA | AK009082.1 | - | 2 - 3 | 322 | 74 | |
AK031497.1 | - | 2 - 3 | 321 | 74 | ||
AK049465.1 | - | 2 - 3 | 330 | 74 | ||
AK144656.1 | - | 2 - 3 | 330 | 74 | ||
AK150985.1 | - | 2 - 3 | 330 | 74 | ||
BC051936.1 | - | 1 - 2 | 65 | 74 | ||
BC083159.1 | - | 1 - 2 | 133 | 74 | ||
BC116437.2 | - | 2 - 3 | 322 | 40104493 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
