Assay Details
| Gene Symbol: Tgfbrap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013025.2 | NP_001013043.1 | 6 - 7 | 1684 | 85 | |
| GenBank mRNA | AK141742.1 | - | 5 - 6 | 1240 | 85 | |
| BC072608.1 | - | 6 - 7 | 1684 | 85 | ||
| BC138866.1 | - | 6 - 7 | 1598 | 8861888 | 85 | |
| BC138867.1 | - | 6 - 7 | 1598 | 8861889 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tgfbrap1
Genomic Map