Assay Details
Gene Symbol: Taf4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100449.1 | NP_001093919.1 | 13 - 14 | 2652 | 66 | |
GenBank mRNA | AK029943.1 | - | 2 - 3 | 197 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
