Assay Details
| Gene Symbol: Mdh1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029696.4 | NP_083972.3 | 6 - 7 | 1078 | 97 | |
| GenBank mRNA | AK007262.1 | - | 2 - 3 | 203 | 97 | |
| BC050786.1 | - | 6 - 7 | 1078 | 6703279 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mdh1b
Genomic Map