Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100019274.mm.1 |
NM_001037742.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019274.mm.1 |
NM_001037742.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019274.mm.1 |
TC0100002371.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019270.mm.1 |
NM_001037742.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019270.mm.1 |
NM_001037742.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019270.mm.1 |
TC0100002371.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019272.mm.1 |
NM_001037742.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019272.mm.1 |
NM_001037742.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019272.mm.1 |
TC0100002371.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019268.mm.1 |
NM_001037742.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019268.mm.1 |
NM_001037742.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019268.mm.1 |
TC0100002371.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019271.mm.1 |
NM_001037742.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019271.mm.1 |
NM_001037742.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019271.mm.1 |
TC0100002371.mm.1 |
MTA TC |