Assay Details
Gene Symbol: Arpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029711.1 | NP_083987.1 | 6 - 7 | 820 | 93 | |
GenBank mRNA | AK008777.1 | - | 2 - 3 | 266 | 93 | |
AK083263.1 | - | 6 - 7 | 630 | 93 | ||
AK151513.1 | - | 6 - 7 | 794 | 93 | ||
AK152356.1 | - | 7 - 8 | 645 | 93 | ||
AK170061.1 | - | 7 - 8 | 619 | 93 | ||
BC099504.1 | - | 5 - 6 | 472 | 93 | ||
BC115750.1 | - | 6 - 7 | 618 | 40051600 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
