Assay Details
Gene Symbol: Arpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029711.1 | NP_083987.1 | 7 - 8 | 953 | 64 | |
GenBank mRNA | AK008777.1 | - | 3 - 4 | 399 | 64 | |
AK083263.1 | - | 7 - 8 | 763 | 64 | ||
AK151513.1 | - | 7 - 8 | 927 | 64 | ||
AK152356.1 | - | 8 - 9 | 778 | 64 | ||
AK170061.1 | - | 8 - 9 | 752 | 64 | ||
BC099504.1 | - | 6 - 7 | 605 | 64 | ||
BC115750.1 | - | 7 - 8 | 751 | 40051600 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
