Assay Details
Gene Symbol: Ino80d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114609.1 | NP_001108081.1 | 3 - 4 | 259 | 72 | |
GenBank mRNA | BC010584.1 | - | 3 - 4 | 327 | 72 | |
BQ030826.1 | - | 2 - 3 | 72 | |||
BY278053.1 | - | 3 - 4 | 259 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001114609.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
