Assay Details
| Gene Symbol: Hibch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146108.1 | NP_666220.1 | 2 - 3 | 144 | 69 | |
| GenBank mRNA | AK076038.1 | - | 2 - 3 | 143 | 69 | |
| AK082965.1 | - | 2 - 3 | 143 | 69 | ||
| AK184752.1 | - | 1 - 2 | 59 | 69 | ||
| AK216816.1 | - | 2 - 3 | 191 | 69 | ||
| BC026437.1 | - | 2 - 3 | 144 | 4238681 | 69 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hibch
Genomic Map