Assay Details
| Gene Symbol: Hibch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146108.1 | NP_666220.1 | 4 - 5 | 368 | 146 | |
| GenBank mRNA | AK076038.1 | - | 4 - 5 | 367 | 146 | |
| AK082965.1 | - | 4 - 5 | 367 | 146 | ||
| AK186066.1 | - | 1 - 2 | 41 | 146 | ||
| BC026437.1 | - | 4 - 5 | 368 | 4238681 | 146 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hibch
Genomic Map