Assay Details
| Gene Symbol: Gnpda1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011937.2 | NP_036067.2 | 6 - 7 | 864 | 75 | |
| GenBank mRNA | AF088903.1 | - | 6 - 7 | 822 | 75 | |
| AF160355.1 | - | 6 - 7 | 810 | 75 | ||
| AK077625.1 | - | 3 - 4 | 529 | 75 | ||
| AK080610.1 | - | 6 - 7 | 813 | 75 | ||
| AK166069.1 | - | 6 - 7 | 815 | 75 | ||
| AK167088.1 | - | 3 - 4 | 538 | 75 | ||
| AK172879.1 | - | 5 - 6 | 727 | 75 | ||
| BC014800.1 | - | 6 - 7 | 795 | 4220980 | 75 | |
| Gene Symbol: Gm8615 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028061.1 | 1 | 647 | 75 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
fructose 6-phosphate metabolic process
glucosamine metabolic process
glucosamine catabolic process
N-acetylglucosamine metabolic process
generation of precursor metabolites and energy
acrosome reaction
fructose biosynthetic process
glucosamine-6-phosphate deaminase activity
hydrolase activity
carbohydrate metabolic process
fructose 6-phosphate metabolic process
glucosamine metabolic process
glucosamine catabolic process
N-acetylglucosamine metabolic process
generation of precursor metabolites and energy
acrosome reaction
fructose biosynthetic process
glucosamine-6-phosphate deaminase activity
hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31633470 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31633471 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31633473 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003658 | CB237974.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003658 | CK338473.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003658 | NR_028061.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003658 | AA839536.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003658 | BE631531.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965892 | NR_028061.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965892 | BF318898.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965892 | BY443103.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965892 | AK166069.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965892 | AK167088.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 70067 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909926 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193973 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 84580 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593029 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494466 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 43450 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119296 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127241 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145981 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003658 | CB237974.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003658 | CK338473.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003658 | NR_028061.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003658 | AA839536.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003658 | BE631531.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965892 | NR_028061.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965892 | BF318898.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965892 | BY443103.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965892 | AK166069.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965892 | AK167088.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P370315 | NR_028061.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P370315 | BE631531.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P370315 | AK166069.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P370315 | AK167088.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P370315 | AK172879.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633471 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31633468 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145981 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119296 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127241 | - | - |
| Whole Mouse Genome G4122A | A_51_P370315 | NR_028061.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P370315 | BE631531.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P370315 | AK166069.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P370315 | AK167088.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P370315 | AK172879.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140055 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121493 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114118 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119296 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145981 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127241 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13798186 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13798199 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13798198 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13798192 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13798194 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31633471 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31633473 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31633470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31633475 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633466 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13670981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633489 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13931731 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633437 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13593725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633505 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633501 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633430 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13777698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633464 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633473 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633461 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633475 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13670983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633512 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633470 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633483 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13798199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633503 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14099283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14111285 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13775488 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31633508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14111269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13931729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14111268 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10458553 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10458551 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10458548 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10458552 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10458554 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535899 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10458550 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10458549 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448163_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10535898 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10458547 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10458547 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10535898 | - | - |
| Mouse Genome 430 2.0 Array | 1448163_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448163_at | - | - |
| Mouse Genome 430 2.0 Array | 1455065_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1455065_PM_x_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500033550.mm.1 | TC0500003651.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500033550.mm.1 | NR_028061.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800009150.mm.1 | NM_011937.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800009150.mm.1 | NM_011937.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800009150.mm.1 | TC1800001230.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800009151.mm.1 | NM_011937.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800009151.mm.1 | NM_011937.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800009151.mm.1 | TC1800001230.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448163_PM_at | - | - |
| Mouse Expression Set 430 | 1455065_PM_x_at | - | - |
| Mouse Expression Set 430 | 1448163_at | - | - |
| Mouse Expression Set 430 | 1455065_x_at | - | - |
| Murine Genome U74v2 Set | 100565_PM_at | - | - |
| Murine Genome U74v2 Set | 100565_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455065_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448163_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455065_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448163_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448163_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455065_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455065_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448163_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 810820 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 840731 | - | - |
| MouseWG-6 v2 Expression BeadChip | 5220112 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 580181 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5220112 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000580181 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005220112 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1228139 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2601063 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2601063 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228139 | - | - |
| mouse-6 v1.1 (Illumina) | scl026384.8_46-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0026384.1_20-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5220112 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 580181 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl026384.8_46-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0026384.1_20-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 580181 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5220112 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1228139 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2750548 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2852217 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1228139 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2852217 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2750548 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0026384.1_20-S | - | - |

Gnpda1
Genomic Map