Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100003937.mm.1 |
NM_001198570.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003937.mm.1 |
NM_001198570.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003937.mm.1 |
NM_001198571.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003937.mm.1 |
NM_001198571.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003937.mm.1 |
NM_198127.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003937.mm.1 |
NM_198127.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003937.mm.1 |
TC0100000471.mm.1 |
MTA TC |