Assay Details
Gene Symbol: Cyp20a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030013.2 | NP_084289.1 | 2 - 3 | 131 | 86 | |
GenBank mRNA | AK020848.1 | - | 2 - 3 | 182 | 86 | |
AK053388.1 | - | 2 - 3 | 134 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
