Assay Details
Gene Symbol: Cyp20a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030013.2 | NP_084289.1 | 4 - 5 | 447 | 86 | |
GenBank mRNA | AK020848.1 | - | 4 - 5 | 498 | 86 | |
AK053388.1 | - | 4 - 5 | 450 | 86 | ||
BC049147.1 | - | 3 - 4 | 264 | 6306072 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
