Assay Details
| Gene Symbol: Dennd5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177192.3 | NP_796166.2 | 15 - 16 | 3317 | 74 | |
| GenBank mRNA | CN537548.1 | - | 3 - 4 | 314 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dennd5b
Genomic Map