Assay Details
| Gene Symbol: Dennd5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177192.3 | NP_796166.2 | 17 - 18 | 3629 | 91 | |
| GenBank mRNA | AK135015.1 | - | 17 - 18 | 3512 | 91 | |
| CV558497.1 | - | 2 - 3 | 214 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dennd5b
Genomic Map