Assay Details
| Gene Symbol: Dennd5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177192.3 | NP_796166.2 | 5 - 6 | 1993 | 62 | |
| GenBank mRNA | AK038858.1 | - | 5 - 6 | 1894 | 62 | |
| AK050728.1 | - | 5 - 6 | 1851 | 62 | ||
| AK135015.1 | - | 5 - 6 | 1878 | 62 | ||
| BC132199.1 | - | 5 - 6 | 1698 | 40130476 | 62 | |
| BC132535.1 | - | 5 - 6 | 1698 | 40130812 | 62 | |
| BC145128.1 | - | 5 - 6 | 1698 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dennd5b
Genomic Map