Assay Details
| Gene Symbol: Tm7sf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK171111.1 | - | 2 - 3 | 343 | 72 | |
| BC023389.1 | - | 2 - 3 | 304 | 72 | ||
| DQ104329.1 | - | 2 - 3 | 314 | 72 | ||
More Information
Set Membership: |
|

Tm7sf3
Genomic Map