Assay Details
| Gene Symbol: Tm7sf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK171111.1 | - | 6 - 7 | 75 | ||
| BC023389.1 | - | 6 - 7 | 922 | 75 | ||
| BC049960.1 | - | 4 - 5 | 75 | |||
| BC057900.1 | - | 1 - 2 | 85 | 75 | ||
| DQ104329.1 | - | 6 - 7 | 931 | 75 | ||
More Information
Set Membership: |
|

Tm7sf3
Genomic Map