Assay Details
| Gene Symbol: Fgfr1op2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026218.2 | NP_080494.1 | 3 - 4 | 86 | ||
| GenBank mRNA | AK004662.1 | - | 3 - 4 | 86 | ||
| AK005320.1 | - | 3 - 4 | 86 | |||
| AK008983.1 | - | 3 - 4 | 86 | |||
| AK050102.1 | - | 3 - 4 | 558 | 87 | ||
| AK151318.1 | - | 3 - 4 | 86 | |||
| AK152617.1 | - | 3 - 4 | 86 | |||
| AK153137.1 | - | 3 - 4 | 86 | |||
| AK169380.1 | - | 3 - 4 | 86 | |||
| AK169383.1 | - | 3 - 4 | 86 | |||
| BC028876.1 | - | 3 - 4 | 4504364 | 86 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fgfr1op2
Genomic Map