Assay Details
| Gene Symbol: Fgfr1op2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026218.2 | NP_080494.1 | 4 - 5 | 699 | 92 | |
| GenBank mRNA | AK004662.1 | - | 4 - 5 | 683 | 92 | |
| AK005320.1 | - | 4 - 5 | 676 | 92 | ||
| AK050102.1 | - | 4 - 5 | 700 | 92 | ||
More Information
Set Membership: |
|

Fgfr1op2
Genomic Map