Assay Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023617.2 | NP_076106.2 | 11 - 12 | 1177 | 67 | |
| GenBank mRNA | AF172276.2 | - | 11 - 12 | 1253 | 67 | |
| AK030109.1 | - | 7 - 8 | 717 | 67 | ||
| AK149434.1 | - | 11 - 12 | 1138 | 67 | ||
| AY347569.1 | - | 11 - 12 | 1054 | 67 | ||
| AY347570.1 | - | 11 - 12 | 1054 | 67 | ||
| BC049853.1 | - | 11 - 12 | 1129 | 67 | ||
| BC138876.1 | - | 11 - 12 | 1156 | 8861898 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aox3
Genomic Map