Assay Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023617.2 | NP_076106.2 | 29 - 30 | 3486 | 71 | |
| GenBank mRNA | AK004700.1 | - | 18 - 19 | 2282 | 71 | |
| AK030109.1 | - | 25 - 26 | 3026 | 72 | ||
| AK149434.1 | - | 29 - 30 | 3448 | 71 | ||
| BC049853.1 | - | 29 - 30 | 3438 | 71 | ||
| BC138876.1 | - | 29 - 30 | 3465 | 8861898 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aox3
Genomic Map