Assay Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023617.2 | NP_076106.2 | 31 - 32 | 3647 | 108 | |
| GenBank mRNA | AF172276.2 | - | 31 - 32 | 3726 | 108 | |
| AK004700.1 | - | 20 - 21 | 2443 | 108 | ||
| AK030109.1 | - | 27 - 28 | 3188 | 108 | ||
| AK149434.1 | - | 31 - 32 | 3609 | 108 | ||
| AY347570.1 | - | 31 - 32 | 3527 | 108 | ||
| BC049853.1 | - | 31 - 32 | 3599 | 108 | ||
| BC138876.1 | - | 31 - 32 | 3626 | 8861898 | 108 | |
More Information
Set Membership: |
|

Aox3
Genomic Map